Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SRRM3
Entrez Gene ID
222183
Gene Name
serine/arginine repetitive matrix 3
Gene Aliases
-
Chromosome Location
Chr.7: 76201893 - 76287292 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001110199.2 NP_001103669.1 15 - 15 3144 141
NM_001291831.1 NP_001278760.1 16 - 16 3223 141
GenBank mRNA AK092590.1 - 3 - 3 1562 141
AK094397.1 - 1 - 1 1755 141
BC063788.1 - 1 - 1 1483 141
BC096755.1 - 4 - 4 1673 141

View Full Product Details