Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
IQCK
Entrez Gene ID
124152
Gene Name
IQ motif containing K
Gene Aliases
-
Chromosome Location
Chr.16: 19716456 - 19858467 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001305121.1 NP_001292050.1 5 - 6 1162 100
NM_001305122.1 NP_001292051.1 6 - 7 1227 100
NM_153208.2 NP_694940.1 6 - 7 1227 100
NR_130968.1 5 - 6 1129 100
XM_011545736.2 XP_011544038.1 5 - 6 562 100
XM_017022933.1 XP_016878422.1 5 - 6 1273 100
GenBank mRNA AF520570.1 - 6 - 7 1227 100
AK301933.1 - 5 - 6 550 100
AK303998.1 - 4 - 5 455 100
AK311216.1 - 4 - 5 422 100
AK315672.1 - 5 - 6 550 100
BC034823.1 - 5 - 6 558 100

View Full Product Details