Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IQCK
Entrez Gene ID
124152
Gene Name
IQ motif containing K
Gene Aliases
-
Chromosome Location
Chr.16: 19716456 - 19858467 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001305121.1 NP_001292050.1 6 - 7 1240 138
NM_153208.2 NP_694940.1 7 - 8 1305 138
NR_130968.1 6 - 7 1207 138
XM_011545736.2 XP_011544038.1 6 - 7 640 138
XM_017022933.1 XP_016878422.1 6 - 7 1351 138
GenBank mRNA AF520570.1 - 7 - 8 1305 138
AK000122.1 - 5 - 6 1374 272
AK303998.1 - 5 - 6 533 138
AK311216.1 - 5 - 6 500 138
AK315672.1 - 6 - 7 628 138
BC034823.1 - 6 - 7 636 138

View Full Product Details