Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ANKFN1
Entrez Gene ID
162282
Gene Name
ankyrin repeat and fibronectin type III domain containing 1
Gene Aliases
-
Chromosome Location
Chr.17: 56110935 - 56517016 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_153228.2 NP_694960.2 16 - 17 2139 94
XM_006721728.3 XP_006721791.1 15 - 16 2362 94
XM_011524428.2 XP_011522730.1 16 - 17 2139 94
XM_011524429.2 XP_011522731.1 16 - 17 2296 94
XM_011524430.2 XP_011522732.1 17 - 18 2262 94
XM_011524431.2 XP_011522733.1 16 - 17 2194 94
XM_011524433.2 XP_011522735.1 10 - 11 1552 94
XM_011524434.2 XP_011522736.1 7 - 8 999 94
XM_017024263.1 XP_016879752.1 18 - 19 2356 94
XM_017024264.1 XP_016879753.1 18 - 19 3341 94
XM_017024265.1 XP_016879754.1 18 - 19 2550 94
XM_017024266.1 XP_016879755.1 16 - 17 2311 94
XM_017024267.1 XP_016879756.1 15 - 16 2163 94
XM_017024268.1 XP_016879757.1 17 - 18 3247 94
XM_017024269.1 XP_016879758.1 15 - 16 2436 94
XM_017024270.1 XP_016879759.1 18 - 19 2354 94
GenBank mRNA AK095654.1 - 16 - 17 2139 94

View Full Product Details