The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001146261.2 | NP_001139733.1 | 7 - 8 | 1373 | 72 |
NM_001146262.2 | NP_001139734.1 | 6 - 7 | 1252 | 72 | |
NM_001146264.2 | NP_001139736.1 | 7 - 8 | 1373 | 72 | |
NM_001256006.1 | NP_001242935.1 | 7 - 8 | 1457 | 72 | |
NM_153262.3 | NP_694994.2 | 6 - 7 | 1252 | 72 | |
XM_006711262.3 | XP_006711325.1 | 5 - 6 | 2431 | 72 | |
XM_017000931.1 | XP_016856420.1 | 5 - 6 | 2431 | 72 | |
XM_017000932.1 | XP_016856421.1 | 7 - 8 | 1447 | 72 | |
XM_017000933.1 | XP_016856422.1 | 7 - 8 | 1310 | 72 | |
XM_017000934.1 | XP_016856423.1 | 4 - 5 | 1106 | 72 | |
XM_017000935.1 | XP_016856424.1 | 5 - 6 | 989 | 72 | |
GenBank mRNA | AB102948.1 | - | 5 - 6 | 1161 | 72 |
AJ617623.1 | - | 6 - 7 | 1195 | 72 | |
AJ617624.1 | - | 6 - 7 | 1338 | 72 | |
AJ617626.1 | - | 3 - 4 | 928 | 72 | |
AJ617627.1 | - | 2 - 3 | 687 | 72 | |
AK091517.1 | - | 6 - 7 | 1175 | 72 | |
BC130323.1 | - | 7 - 8 | 1306 | 72 | |
BC144154.1 | - | 7 - 8 | 1306 | 72 | |
BC144155.1 | - | 6 - 7 | 1185 | 72 | |
BC144157.1 | - | 7 - 8 | 1306 | 72 |