The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_153362.2 | NP_699193.2 | 1 - 2 | 217 | 102 |
GenBank mRNA | AK291928.1 | - | 1 - 2 | 102 | 102 |
BC037170.1 | - | 1 - 2 | 140 | 102 | |
DA251169.1 | - | 1 - 2 | 217 | 102 |