The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_027064.2 | 2 - 3 | 501 | 92 | |
GenBank mRNA | BC036545.1 | - | 2 - 3 | 468 | 92 |
BC120980.1 | - | 1 - 2 | 114 | 92 | |
BC120981.1 | - | 1 - 2 | 114 | 92 | |
DA059417.1 | - | 4 - 5 | 406 | 92 |