Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KLHDC1
Entrez Gene ID
122773
Gene Name
kelch domain containing 1
Gene Aliases
MST025
Chromosome Location
Chr.14: 49693105 - 49753152 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172193.2 NP_751943.1 2 - 3 252 141
XM_011536422.2 XP_011534724.1 2 - 3 175 141
GenBank mRNA AF111806.1 - 2 - 3 190 141
AK127202.1 - 2 - 3 175 141
BC031270.1 - 2 - 3 215 141
BC101595.1 - 2 - 3 252 141
BC101597.1 - 2 - 3 252 141

View Full Product Details