Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KLHDC1
Entrez Gene ID
122773
Gene Name
kelch domain containing 1
Gene Aliases
MST025
Chromosome Location
Chr.14: 49693105 - 49753152 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172193.2 NP_751943.1 4 - 5 488 77
XM_011536422.2 XP_011534724.1 4 - 5 411 77
GenBank mRNA AF111806.1 - 4 - 5 426 77
AK127202.1 - 4 - 5 411 77
BC101595.1 - 4 - 5 488 77
BC101597.1 - 4 - 5 488 77

View Full Product Details