The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001278669.1 | NP_001265598.1 | 4 - 5 | 2043 | 80 |
NM_001278670.1 | NP_001265599.1 | 4 - 5 | 2043 | 80 | |
NM_001278672.1 | NP_001265601.1 | 4 - 5 | 1669 | 80 | |
NM_001278673.1 | NP_001265602.1 | 3 - 4 | 570 | 80 | |
NM_001278675.1 | NP_001265604.1 | 4 - 5 | 1669 | 80 | |
NM_006162.4 | NP_006153.2 | 4 - 5 | 2043 | 80 | |
NM_172387.2 | NP_765975.1 | 4 - 5 | 1669 | 80 | |
NM_172388.2 | NP_765976.1 | 3 - 4 | 944 | 80 | |
NM_172389.2 | NP_765977.1 | 4 - 5 | 1669 | 80 | |
NM_172390.2 | NP_765978.1 | 4 - 5 | 2043 | 80 | |
XM_017025783.1 | XP_016881272.1 | 4 - 5 | 1885 | 80 | |
GenBank mRNA | AK292641.1 | - | 4 - 5 | 1885 | 80 |
AK293760.1 | - | 3 - 4 | 515 | 80 | |
BC104753.1 | - | 4 - 5 | 1684 | 80 | |
BC112243.1 | - | 4 - 5 | 1684 | 80 | |
BC143457.1 | - | 4 - 5 | 1684 | 80 | |
EU887559.1 | - | 4 - 5 | 1590 | 80 | |
EU887560.1 | - | 4 - 5 | 1551 | 80 | |
EU887561.1 | - | 4 - 5 | 1590 | 80 | |
EU887562.1 | - | 4 - 5 | 1551 | 80 | |
EU887563.1 | - | 4 - 5 | 1590 | 80 | |
EU887564.1 | - | 4 - 5 | 1551 | 80 | |
EU887565.1 | - | 4 - 5 | 1590 | 80 | |
EU887566.1 | - | 4 - 5 | 1551 | 80 | |
U08015.1 | - | 4 - 5 | 1829 | 80 | |
U59736.1 | - | 4 - 5 | 1626 | 80 | |
U80917.1 | - | 4 - 5 | 1959 | 80 | |
U80918.1 | - | 4 - 5 | 1613 | 80 | |
U80919.1 | - | 3 - 4 | 772 | 80 |