Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CPO
Entrez Gene ID
130749
Gene Name
carboxypeptidase O
Gene Aliases
-
Chromosome Location
Chr.2: 206932853 - 206971064 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_173077.2 NP_775100.1 6 - 7 620 82
XM_017003372.1 XP_016858861.1 7 - 8 6727 82
GenBank mRNA AJ422118.1 - 6 - 7 574 82
BC112076.1 - 6 - 7 82
BC112078.1 - 6 - 7 82

View Full Product Details