Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MISP
Entrez Gene ID
126353
Gene Name
mitotic spindle positioning
Gene Aliases
C19orf21, MISP1
Chromosome Location
Chr.19: 748413 - 764319 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_173481.3 NP_775752.1 2 - 3 1896 126
XM_011527685.2 XP_011525987.1 2 - 3 1941 126
XM_011527686.2 XP_011525988.1 2 - 3 2034 126
GenBank mRNA BC042125.2 - 2 - 3 1863 126
BC052236.1 - 2 - 3 1883 126
BX648389.1 - 2 - 3 1877 126
BX648815.1 - 1 - 2 1084 126

View Full Product Details