The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001300845.1 | NP_001287774.1 | 2 - 3 | 688 | 79 |
NM_173508.3 | NP_775779.1 | 3 - 4 | 973 | 79 | |
GenBank mRNA | AK095031.1 | - | 3 - 4 | 754 | 79 |
BC037878.1 | - | 2 - 3 | 632 | 79 |