The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_173519.2 | NP_775790.1 | 3 - 4 | 953 | 60 |
XM_017013141.1 | XP_016868630.1 | 4 - 5 | 944 | 60 | |
XM_017013142.1 | XP_016868631.1 | 4 - 5 | 1101 | 60 | |
GenBank mRNA | AF445194.1 | - | 3 - 4 | 947 | 60 |
AK125153.1 | - | 2 - 3 | 200 | 60 | |
AK313040.1 | - | 3 - 4 | 900 | 60 | |
AY094971.1 | - | 3 - 4 | 911 | 60 | |
BC042617.1 | - | 3 - 4 | 928 | 60 |