The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020945.1 | NP_065996.1 | 9 - 10 | 1654 | 61 |
XM_005270004.3 | XP_005270061.1 | 9 - 10 | 1769 | 61 | |
XM_011539986.2 | XP_011538288.2 | 9 - 10 | 1772 | 61 | |
XM_011539987.2 | XP_011538289.2 | 9 - 10 | 1775 | 61 | |
XM_011539988.2 | XP_011538290.1 | 10 - 11 | 1853 | 61 | |
XM_011539990.2 | XP_011538292.1 | 9 - 10 | 1800 | 61 | |
XM_011539991.2 | XP_011538293.1 | 9 - 10 | 1804 | 61 | |
XM_011539992.2 | XP_011538294.1 | 9 - 10 | 1804 | 61 | |
XM_017016463.1 | XP_016871952.1 | 10 - 11 | 1696 | 61 | |
XM_017016465.1 | XP_016871954.1 | 9 - 10 | 1781 | 61 | |
GenBank mRNA | AK127650.1 | - | 9 - 10 | 1663 | 61 |
BC034937.1 | - | 9 - 10 | 1720 | 61 |