Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FNDC7
Entrez Gene ID
163479
Gene Name
fibronectin type III domain containing 7
Gene Aliases
-
Chromosome Location
Chr.1: 108712934 - 108742749 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001144937.1 NP_001138409.1 6 - 7 1112 114
GenBank mRNA AK093157.1 - 2 - 3 438 114
BC057762.1 - 2 - 3 485 114
BC130421.1 - 2 - 3 438 114

View Full Product Details