Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TRIML2
Entrez Gene ID
205860
Gene Name
tripartite motif family like 2
Gene Aliases
SPRYD6
Chromosome Location
Chr.4: 188091255 - 188109603 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001303419.1 NP_001290348.1 2 - 3 505 130
NM_173553.2 NP_775824.2 2 - 3 505 130
XM_011531737.2 XP_011530039.1 3 - 4 696 130
XM_011531738.1 XP_011530040.1 3 - 4 866 130
XM_017007882.1 XP_016863371.1 3 - 4 551 130
GenBank mRNA AK098667.1 - 2 - 3 172 130
AK300428.1 - 3 - 4 534 130
BC111959.1 - 2 - 3 166 130
BC111961.1 - 2 - 3 166 130
BC143709.1 - 2 - 3 166 130

View Full Product Details