Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZDHHC23
Entrez Gene ID
254887
Gene Name
zinc finger DHHC-type containing 23
Gene Aliases
DHHC-23, NIDD
Chromosome Location
Chr.3: 113947901 - 113998002 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001320466.1 NP_001307395.1 3 - 4 1279 102
NM_001320467.1 NP_001307396.1 3 - 4 1171 102
NM_173570.4 NP_775841.2 3 - 4 1171 102
NR_135271.1 3 - 4 1279 102
XM_005247269.1 XP_005247326.1 3 - 4 1115 102
XM_005247270.3 XP_005247327.1 2 - 3 410 102
XM_006713562.3 XP_006713625.1 3 - 4 1115 102
XM_011512618.2 XP_011510920.1 3 - 4 1119 102
XM_011512619.1 XP_011510921.1 3 - 4 1115 102
XM_017006085.1 XP_016861574.1 3 - 4 1094 102
GenBank mRNA AK127025.1 - 3 - 4 1115 102
AK127996.1 - 4 - 5 1116 102
BC035230.1 - 3 - 4 1171 102

View Full Product Details