The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001271983.1 | NP_001258912.1 | 1 - 2 | 235 | 102 |
GenBank mRNA | AK096377.1 | - | 1 - 2 | 235 | 102 |