The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_173595.3 | NP_775866.2 | 17 - 18 | 1870 | 61 |
XM_011538197.2 | XP_011536499.1 | 16 - 17 | 1972 | 61 | |
XM_017019183.1 | XP_016874672.1 | 16 - 17 | 2053 | 61 | |
XM_017019184.1 | XP_016874673.1 | 17 - 18 | 1902 | 61 | |
GenBank mRNA | AB190990.1 | - | 17 - 18 | 1887 | 61 |
AK125379.1 | - | 17 - 18 | 1887 | 61 | |
AK126893.1 | - | 10 - 11 | 1030 | 61 |