Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C2orf61
Entrez Gene ID
285051
Gene Name
chromosome 2 open reading frame 61
Gene Aliases
-
Chromosome Location
Chr.2: 47086991 - 47155378 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001163561.1 NP_001157033.1 1 - 2 209 113
NM_173649.2 NP_775920.1 1 - 2 209 113
GenBank mRNA AK097491.1 - 1 - 2 118 113
DA313162.1 - 1 - 2 209 113

View Full Product Details