The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001012761.2 | NP_001012779.2 | 2 - 3 | 408 | 154 |
XM_011543345.2 | XP_011541647.1 | 2 - 3 | 169 | 154 | |
XM_011543347.2 | XP_011541649.2 | 1 - 2 | 92 | 154 | |
XM_017009390.1 | XP_016864879.1 | 2 - 3 | 656 | 154 | |
XM_017009391.1 | XP_016864880.1 | 2 - 3 | 554 | 154 | |
GenBank mRNA | AK074887.1 | - | 2 - 3 | 655 | 154 |
BC067736.1 | - | 2 - 3 | 408 | 154 |