The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_173674.2 | NP_775945.1 | 12 - 13 | 1622 | 74 |
XM_005266941.1 | XP_005266998.1 | 12 - 13 | 1586 | 74 | |
XM_011535771.1 | XP_011534073.1 | 13 - 14 | 1671 | 74 | |
XM_011535773.2 | XP_011534075.1 | 12 - 13 | 1586 | 74 | |
XM_017010775.1 | XP_016866264.1 | 13 - 14 | 1595 | 74 | |
GenBank mRNA | AK098194.1 | - | 4 - 5 | 414 | 74 |
BC035671.1 | - | 12 - 13 | 1569 | 74 | |
CR749448.1 | - | 8 - 9 | 3174 | 74 |