The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145716.2 | NP_001139188.1 | 6 - 7 | 1299 | 75 |
NM_001145717.1 | NP_001139189.2 | 6 - 7 | 1383 | 75 | |
NM_173676.2 | NP_775947.2 | 6 - 7 | 1272 | 75 | |
XM_011514519.2 | XP_011512821.1 | 6 - 7 | 2016 | 75 | |
XM_011514520.2 | XP_011512822.1 | 6 - 7 | 1917 | 75 | |
XM_017010776.1 | XP_016866265.1 | 6 - 7 | 2046 | 75 | |
XM_017010777.1 | XP_016866266.1 | 6 - 7 | 2045 | 75 | |
XM_017010778.1 | XP_016866267.1 | 6 - 7 | 1944 | 75 | |
XM_017010779.1 | XP_016866268.1 | 5 - 6 | 979 | 75 | |
GenBank mRNA | AK096074.1 | - | 6 - 7 | 1272 | 75 |
AM182887.1 | - | 6 - 7 | 1386 | 75 | |
BC103905.1 | - | 6 - 7 | 1264 | 75 | |
BC103906.1 | - | 6 - 7 | 1237 | 75 | |
BC103907.1 | - | 6 - 7 | 1237 | 75 | |
FJ457781.1 | - | 6 - 7 | 1383 | 75 |