Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HIST1H2BE
Entrez Gene ID
8344
Gene Name
histone cluster 1, H2be
Gene Aliases
H2B.h, H2B/h, H2BFH, dJ221C16.8
Chromosome Location
Chr.6: 26183796 - 26184230 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003523.2 NP_003514.2 1 - 1 352 77
GenBank mRNA BC101748.1 - 1 - 1 419 77
BC101750.1 - 1 - 1 418 77

View Full Product Details