The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003588.3 | NP_003579.3 | 1 - 2 | 418 | 88 |
XM_005262481.1 | XP_005262538.1 | 1 - 2 | 418 | 88 | |
GenBank mRNA | AK315037.1 | - | 1 - 2 | 354 | 88 |
BC036216.1 | - | 1 - 2 | 383 | 88 | |
DB092748.1 | - | 1 - 2 | 418 | 88 |