The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006011.3 | NP_006002.1 | 5 - 6 | 996 | 83 |
XM_017022642.1 | XP_016878131.1 | 5 - 6 | 1159 | 83 | |
XM_017022643.1 | XP_016878132.1 | 4 - 5 | 932 | 83 | |
GenBank mRNA | AK313924.1 | - | 5 - 6 | 944 | 83 |
BC069584.1 | - | 5 - 6 | 865 | 83 | |
BC096202.3 | - | 5 - 6 | 865 | 83 | |
BC096203.3 | - | 5 - 6 | 865 | 83 | |
BC096204.3 | - | 5 - 6 | 865 | 83 | |
BC096205.3 | - | 4 - 5 | 802 | 83 | |
L29556.1 | - | 2 - 3 | 382 | 83 | |
U33551.1 | - | 5 - 6 | 841 | 83 | |
U82762.1 | - | 5 - 6 | 866 | 83 |