Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IFT88
Entrez Gene ID
8100
Gene Name
intraflagellar transport 88
Gene Aliases
D13S1056E, DAF19, TG737, TTC10, hTg737
Chromosome Location
Chr.13: 20566446 - 20691437 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001318491.1 NP_001305420.1 20 - 21 2390 127
NM_001318493.1 NP_001305422.1 22 - 23 2775 127
NM_006531.4 NP_006522.2 21 - 22 2447 127
NM_175605.4 NP_783195.2 23 - 24 2676 127
XM_005266553.2 XP_005266610.1 23 - 24 3037 127
XM_006719870.3 XP_006719933.1 21 - 22 2142 127
XM_011535241.2 XP_011533543.1 22 - 23 2250 127
XM_011535242.1 XP_011533544.1 24 - 25 3177 127
XM_011535243.1 XP_011533545.1 23 - 24 3056 127
XM_017020757.1 XP_016876246.1 22 - 23 3155 127
XM_017020758.1 XP_016876247.1 21 - 22 2651 127
XM_017020759.1 XP_016876248.1 21 - 22 2621 127
XM_017020760.1 XP_016876249.1 24 - 25 2507 127
XM_017020761.1 XP_016876250.1 23 - 24 2143 127
XM_017020762.1 XP_016876251.1 22 - 23 2708 127
XM_017020764.1 XP_016876253.1 22 - 23 2052 127
XM_017020765.1 XP_016876254.1 21 - 22 2154 127
XM_017020766.1 XP_016876255.1 19 - 20 1978 127
XM_017020767.1 XP_016876256.1 21 - 22 1974 127
XM_017020768.1 XP_016876257.1 22 - 23 2519 127
XM_017020770.1 XP_016876259.1 24 - 25 2507 127
XM_017020771.1 XP_016876260.1 24 - 25 2935 127
XM_017020772.1 XP_016876261.1 22 - 23 2052 127
XM_017020774.1 XP_016876263.1 20 - 21 2030 127
GenBank mRNA AK300769.1 - 20 - 21 2054 127
AK308281.1 - 21 - 22 2102 127
BC030776.1 - 23 - 24 2357 127
U20362.1 - 21 - 22 2191 127

View Full Product Details