Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CHL1
Entrez Gene ID
10752
Gene Name
cell adhesion molecule L1 like
Gene Aliases
CALL, L1CAM2
Chromosome Location
Chr.3: 196588 - 409417 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001253387.1 NP_001240316.1 6 - 7 1155 121
NM_001253388.1 NP_001240317.1 4 - 5 607 121
NM_006614.3 NP_006605.2 6 - 7 1155 121
XM_006712938.1 XP_006713001.1 6 - 7 766 121
XM_006712939.3 XP_006713002.1 5 - 6 1083 121
XM_006712940.3 XP_006713003.1 5 - 6 997 121
XM_011533292.1 XP_011531594.1 6 - 7 1136 121
XM_011533294.1 XP_011531596.1 6 - 7 766 121
XM_011533295.1 XP_011531597.1 6 - 7 1136 121
XM_011533296.1 XP_011531598.1 6 - 7 766 121
XM_017005566.1 XP_016861055.1 7 - 8 5660 121
XM_017005567.1 XP_016861056.1 5 - 6 868 121
XM_017005568.1 XP_016861057.1 5 - 6 675 121
XM_017005569.1 XP_016861058.1 5 - 6 1083 121
XM_017005570.1 XP_016861059.1 5 - 6 997 121
XM_017005571.1 XP_016861060.1 5 - 6 868 121
XM_017005572.1 XP_016861061.1 6 - 7 1163 121
XM_017005573.1 XP_016861062.1 5 - 6 1083 121
GenBank mRNA AB209329.1 - 6 - 7 980 121
AF002246.1 - 6 - 7 784 121
AK126878.1 - 6 - 7 1134 121
BC104918.1 - 4 - 5 544 121
BC143496.1 - 4 - 5 541 121

View Full Product Details