Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 35577146-35577387 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PEX12
Entrez Gene ID
5193
Gene Name
peroxisomal biogenesis factor 12
Gene Aliases
PAF-3, PBD3A
UniGene
Hs.591190
Gene Chromosome Location
Chr.17: 35574795-35578637 on Build GRCh38
Transcripts
NM_000286.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
705496 c.431C>G p.S144C Chr.17: 35577287-35577287 on Build GRCh38
4634188 c.349A>T p.I117F Chr.17: 35577369-35577369 on Build GRCh38
4614799 c.368_370delTTC p.L123delL Chr.17: 35577348-35577350 on Build GRCh38
176755 c.539G>A p.R180Q Chr.17: 35577179-35577179 on Build GRCh38
7117528 c.367C>T p.L123F Chr.17: 35577351-35577351 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768059114 Chr.17: 35577232-35577232 on Build GRCh38
rs757097977 Chr.17: 35577285-35577285 on Build GRCh38
rs773967871 Chr.17: 35577330-35577330 on Build GRCh38
rs17852912 Chr.17: 35577309-35577309 on Build GRCh38
rs769196917 Chr.17: 35577303-35577303 on Build GRCh38
rs946528890 Chr.17: 35577300-35577300 on Build GRCh38
rs768259519 Chr.17: 35577327-35577327 on Build GRCh38
rs956055423 Chr.17: 35577283-35577283 on Build GRCh38
rs762220033 Chr.17: 35577346-35577346 on Build GRCh38
rs1042122765 Chr.17: 35577230-35577230 on Build GRCh38
rs564816937 Chr.17: 35577257-35577257 on Build GRCh38
rs202025715 Chr.17: 35577204-35577204 on Build GRCh38
rs751442386 Chr.17: 35577280-35577280 on Build GRCh38
rs61752103 Chr.17: 35577180-35577180 on Build GRCh38
rs766411953 Chr.17: 35577261-35577261 on Build GRCh38
rs986848130 Chr.17: 35577268-35577268 on Build GRCh38
rs945104524 Chr.17: 35577207-35577207 on Build GRCh38
rs768919252 Chr.17: 35577187-35577187 on Build GRCh38
rs751058068 Chr.17: 35577348-35577350 on Build GRCh38
rs923109489 Chr.17: 35577200-35577200 on Build GRCh38
rs974295574 Chr.17: 35577228-35577228 on Build GRCh38
rs762603832 Chr.17: 35577183-35577183 on Build GRCh38
rs61752102 Chr.17: 35577183-35577185 on Build GRCh38
rs369522860 Chr.17: 35577290-35577290 on Build GRCh38
rs772417571 Chr.17: 35577358-35577358 on Build GRCh38
rs903493307 Chr.17: 35577231-35577231 on Build GRCh38
rs192653658 Chr.17: 35577175-35577175 on Build GRCh38
rs988442642 Chr.17: 35577287-35577287 on Build GRCh38
rs150186509 Chr.17: 35577266-35577266 on Build GRCh38
rs748918869 Chr.17: 35577312-35577312 on Build GRCh38
rs745420734 Chr.17: 35577179-35577179 on Build GRCh38
rs779716641 Chr.17: 35577246-35577247 on Build GRCh38
rs61752104 Chr.17: 35577176-35577177 on Build GRCh38
rs781337837 Chr.17: 35577296-35577296 on Build GRCh38
rs138731505 Chr.17: 35577267-35577267 on Build GRCh38
rs774653352 Chr.17: 35577188-35577188 on Build GRCh38
rs375119247 Chr.17: 35577271-35577271 on Build GRCh38
rs748397220 Chr.17: 35577360-35577360 on Build GRCh38
rs926359179 Chr.17: 35577342-35577342 on Build GRCh38
rs762340754 Chr.17: 35577195-35577195 on Build GRCh38
rs745813377 Chr.17: 35577298-35577298 on Build GRCh38

View Full Product Details