The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018384.4 | NP_060854.2 | 2 - 3 | 404 | 98 |
GenBank mRNA | AK297881.1 | - | 3 - 4 | 228 | 98 |
AL557446.3 | - | 2 - 3 | 178 | 98 | |
BC011732.2 | - | 2 - 3 | 137 | 98 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001199577.1 | NP_001186506.1 | 5 - 6 | 789 | 98 |
NM_001303630.1 | NP_001290559.1 | 4 - 5 | 430 | 98 |