Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BBS1
Entrez Gene ID
582
Gene Name
Bardet-Biedl syndrome 1
Gene Aliases
BBS2L2
Chromosome Location
Chr.11: 66510648 - 66533613 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024649.4 NP_078925.3 7 - 8 600 69
GenBank mRNA AF503941.1 - 7 - 8 600 69
AK095638.1 - 6 - 7 607 69
AK302657.1 - 5 - 6 531 69
BC047642.1 - 6 - 7 559 69
BC109064.2 - 7 - 8 593 69
BC109065.1 - 7 - 8 593 69
BX647612.1 - 7 - 8 629 69

View Full Product Details