Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BBS1
Entrez Gene ID
582
Gene Name
Bardet-Biedl syndrome 1
Gene Aliases
BBS2L2
Chromosome Location
Chr.11: 66510648 - 66533613 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024649.4 NP_078925.3 11 - 12 1119 63
GenBank mRNA AF503941.1 - 11 - 12 1119 63
AK095638.1 - 10 - 11 1126 63
AK294962.1 - 3 - 4 353 63
AK296311.1 - 7 - 8 834 63
BC047642.1 - 10 - 11 1078 63
BC109064.2 - 11 - 12 1112 63
BX647612.1 - 11 - 12 1148 63

View Full Product Details