The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025003.3 | NP_079279.3 | 11 - 12 | 1614 | 82 |
XM_011538754.2 | XP_011537056.1 | 11 - 12 | 1667 | 82 | |
XM_017019979.1 | XP_016875468.1 | 4 - 5 | 683 | 82 | |
GenBank mRNA | AF488804.1 | - | 11 - 12 | 1614 | 82 |
AJ515153.3 | - | 11 - 12 | 1614 | 82 | |
AJ515154.2 | - | 11 - 12 | 1614 | 82 |