The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001252675.1 | NP_001239604.1 | 6 - 7 | 1187 | 122 |
NM_001252676.1 | NP_001239605.1 | 5 - 6 | 1014 | 122 | |
NM_001252677.1 | NP_001239606.1 | 6 - 7 | 1187 | 122 | |
NM_032501.3 | NP_115890.2 | 6 - 7 | 1187 | 122 | |
XM_011529388.1 | XP_011527690.1 | 5 - 6 | 972 | 122 | |
GenBank mRNA | AB058749.1 | - | 1 - 2 | 104 | 122 |
AK092295.1 | - | 4 - 5 | 596 | 122 | |
AK125058.1 | - | 4 - 5 | 2390 | 122 | |
AK127566.1 | - | 6 - 7 | 1133 | 122 | |
AK296306.1 | - | 5 - 6 | 1011 | 122 | |
AK314440.1 | - | 6 - 7 | 1110 | 122 | |
BC039261.1 | - | 6 - 7 | 1146 | 122 | |
BC044588.1 | - | 6 - 7 | 1112 | 122 | |
BC055008.1 | - | 6 - 7 | 1112 | 122 | |
BC114612.1 | - | 6 - 7 | 978 | 122 |