The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_033103.4 | NP_149094.3 | 8 - 9 | 1012 | 119 |
GenBank mRNA | AF423421.1 | - | 8 - 9 | 946 | 119 |
AJ347750.1 | - | 8 - 9 | 946 | 119 | |
AK098246.1 | - | 8 - 9 | 982 | 119 | |
AK126506.1 | - | 5 - 6 | 1280 | 119 | |
AK300775.1 | - | 7 - 8 | 906 | 119 | |
AK315105.1 | - | 8 - 9 | 982 | 119 | |
AL831950.1 | - | 7 - 8 | 860 | 119 | |
BC036447.2 | - | 8 - 9 | 982 | 119 |