Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SNX3
Entrez Gene ID
8724
Gene Name
sorting nexin 3
Gene Aliases
Grd19, MCOPS8, SDP3
Chromosome Location
Chr.6: 108211217 - 108261260 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC016863.2 - 2 - 3 607 66

View Full Product Details