The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_153486.3 | NP_705690.2 | 3 - 4 | 383 | 60 |
NM_194436.2 | NP_919417.1 | 3 - 4 | 383 | 60 | |
GenBank mRNA | AY092767.1 | - | 3 - 4 | 388 | 60 |
BC040279.1 | - | 3 - 4 | 348 | 60 | |
BC047902.1 | - | 3 - 4 | 378 | 60 | |
BM544922.1 | - | 3 - 4 | 389 | 60 |