The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003856.3 | NP_003847.2 | 3 - 4 | 511 | 111 |
NM_016232.4 | NP_057316.3 | 3 - 4 | 545 | 111 | |
NR_104167.1 | 3 - 4 | 511 | 111 | ||
XM_006712839.3 | XP_006712902.1 | 3 - 4 | 511 | 111 | |
XM_011512151.1 | XP_011510453.1 | 3 - 4 | 518 | 111 | |
GenBank mRNA | AB012701.3 | - | 3 - 4 | 545 | 111 |
AK291578.1 | - | 3 - 4 | 503 | 111 | |
AL553735.3 | - | 3 - 4 | 497 | 111 | |
BC030975.1 | - | 3 - 4 | 504 | 111 | |
D12763.1 | - | 2 - 3 | 320 | 111 | |
DA884586.1 | - | 3 - 4 | 512 | 111 |