Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BCORP1
Entrez Gene ID
286554
Gene Name
BCL6 corepressor pseudogene 1
Gene Aliases
BCORL2
Chromosome Location
Chr.Y: 19455431 - 19503153 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_002923.2 4 - 5 752 100
NR_033732.1 4 - 5 752 100
GenBank mRNA AK097140.1 - 4 - 5 752 100
BC063452.1 - 4 - 5 735 100
BC110520.1 - 3 - 4 627 100
BC110521.1 - 3 - 4 627 100


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream testis-specific transcript, Y-linked 14 (non-protein coding) TTTY14 CYorf14, NCRNA00137, NCRNA00185, PRO2834, TTY14 -388.9kb Sense Chr. - 18872501 - 19077547 on Build GRCh38
Downstream taxilin gamma pseudogene, Y-linked TXLNGY CYorf15A, CYorf15B, TXLNG2P 93.2kb Anti-sense Chr. - 19567358 - 19607170 on Build GRCh38

View Full Product Details