Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CES4A
Entrez Gene ID
283848
Gene Name
carboxylesterase 4A
Gene Aliases
CES6, CES8
Chromosome Location
Chr.16: 66988589 - 67009758 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001190201.1 NP_001177130.1 5 - 6 766 60
NM_001190202.1 NP_001177131.1 5 - 6 778 60
NM_001318506.1 NP_001305435.1 5 - 6 822 60
NM_173815.6 NP_776176.5 7 - 8 1085 60
XM_005255896.4 XP_005255953.1 7 - 8 1082 60
XM_011523021.2 XP_011521323.1 8 - 9 1214 60
XM_011523023.2 XP_011521325.1 8 - 9 1202 60
XM_011523024.2 XP_011521326.1 8 - 9 1214 60
XM_011523025.2 XP_011521327.1 8 - 9 1214 60
XM_011523026.2 XP_011521328.1 7 - 8 1094 60
XM_011523027.2 XP_011521329.1 8 - 9 1214 60
XM_011523028.2 XP_011521330.1 7 - 8 1102 60
XM_011523029.2 XP_011521331.1 8 - 9 1214 60
XM_011523030.2 XP_011521332.1 7 - 8 1082 60
XM_017023160.1 XP_016878649.1 8 - 9 1202 60
XM_017023161.1 XP_016878650.1 8 - 9 1202 60
XM_017023162.1 XP_016878651.1 8 - 9 1214 60
XM_017023163.1 XP_016878652.1 8 - 9 1214 60
XM_017023164.1 XP_016878653.1 8 - 9 1202 60
XM_017023165.1 XP_016878654.1 8 - 9 1202 60
XM_017023166.1 XP_016878655.1 8 - 9 1214 60
XM_017023167.1 XP_016878656.1 5 - 6 1166 60
XM_017023168.1 XP_016878657.1 5 - 6 1172 60
GenBank mRNA AK094783.1 - 5 - 6 783 60
AK293061.1 - 5 - 6 614 60
AK295483.1 - 5 - 6 718 60
AK296064.1 - 7 - 8 1005 60
AK300792.1 - 5 - 6 726 60
BC064573.1 - 5 - 6 546 60
BC084555.1 - 6 - 7 790 60

View Full Product Details