Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CES4A
Entrez Gene ID
283848
Gene Name
carboxylesterase 4A
Gene Aliases
CES6, CES8
Chromosome Location
Chr.16: 66988589 - 67009758 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001190201.1 NP_001177130.1 6 - 7 802 61
NM_001190202.1 NP_001177131.1 6 - 7 814 61
NM_001318506.1 NP_001305435.1 6 - 7 858 61
NM_173815.6 NP_776176.5 8 - 9 1121 61
XM_005255896.4 XP_005255953.1 8 - 9 1118 61
XM_011523021.2 XP_011521323.1 9 - 10 1250 61
XM_011523023.2 XP_011521325.1 9 - 10 1238 61
XM_011523024.2 XP_011521326.1 9 - 10 1250 61
XM_011523025.2 XP_011521327.1 9 - 10 1250 61
XM_011523026.2 XP_011521328.1 8 - 9 1130 61
XM_011523027.2 XP_011521329.1 9 - 10 1250 61
XM_011523028.2 XP_011521330.1 8 - 9 1138 61
XM_011523029.2 XP_011521331.1 9 - 10 1250 61
XM_011523030.2 XP_011521332.1 8 - 9 1118 61
XM_017023160.1 XP_016878649.1 9 - 10 1238 61
XM_017023161.1 XP_016878650.1 9 - 10 1238 61
XM_017023162.1 XP_016878651.1 9 - 10 1250 61
XM_017023163.1 XP_016878652.1 9 - 10 1250 61
XM_017023164.1 XP_016878653.1 9 - 10 1238 61
XM_017023165.1 XP_016878654.1 9 - 10 1238 61
XM_017023166.1 XP_016878655.1 9 - 10 1250 61
XM_017023167.1 XP_016878656.1 6 - 7 1202 61
XM_017023168.1 XP_016878657.1 6 - 7 1208 61
GenBank mRNA AK094783.1 - 6 - 7 819 61
AK293061.1 - 6 - 7 650 61
AK295483.1 - 6 - 7 754 61
AK296064.1 - 8 - 9 1041 61
AK300792.1 - 6 - 7 762 61
BC064573.1 - 6 - 7 582 61
BC084555.1 - 7 - 8 826 61

View Full Product Details