The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_173856.2 | NP_776255.2 | 1 - 1 | 532 | 120 |
GenBank mRNA | BC130356.1 | - | 1 - 1 | 532 | 120 |
BC130358.1 | - | 1 - 1 | 532 | 120 |