Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
VN1R5
Entrez Gene ID
317705
Gene Name
vomeronasal 1 receptor 5 (gene/pseudogene)
Gene Aliases
V1RL5
Chromosome Location
Chr.1: 247256072 - 247257145 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_173858.1 NP_776257.1 1 - 1 161 132
GenBank mRNA BC126341.1 - 1 - 1 261 132
BC126367.1 - 1 - 1 261 132

View Full Product Details