Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BPIFA4P
Entrez Gene ID
317716
Gene Name
BPI fold containing family A member 4, pseudogene
Gene Aliases
BASE
Chromosome Location
Chr.20: 33193605 - 33210462 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_026760.1 1 - 2 38 67
GenBank mRNA AY180924.1 - 1 - 2 38 67
BC069315.1 - 1 - 2 37 67


Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BX642190.1 - 1 - 2 59 Chr. 20 - 33193585 - 33198767 [+] on Build GRCh38 67
GenBankmRNA BX642280.1 - 1 - 2 59 Chr. 20 - 33193585 - 33198796 [+] on Build GRCh38 67


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream BPI fold containing family A member 2 BPIFA2 C20orf70, PSP, SPLUNC2, bA49G10.1 -12.2kb Sense Chr. - 33161848 - 33181417 on Build GRCh38
Downstream BPI fold containing family A member 3 BPIFA3 C20orf71, SPLUNC3 19.0kb Sense Chr. - 33217310 - 33227806 on Build GRCh38

View Full Product Details