Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FAM98C
Entrez Gene ID
147965
Gene Name
family with sequence similarity 98 member C
Gene Aliases
-
Chromosome Location
Chr.19: 38403124 - 38409088 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_174905.3 NP_777565.3 6 - 7 772 100
GenBank mRNA BC034383.1 - 3 - 4 341 100
BC117258.1 - 6 - 7 772 100
BC143830.1 - 6 - 7 772 100
BI871449.1 - 3 - 4 304 100
BM561971.1 - 6 - 7 714 100

View Full Product Details