The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_174916.2 | NP_777576.1 | 4 - 5 | 591 | 106 |
GenBank mRNA | AF525401.1 | - | 4 - 5 | 474 | 106 |
AK056441.1 | - | 4 - 5 | 541 | 106 | |
AK292516.1 | - | 4 - 5 | 538 | 106 | |
AK302489.1 | - | 4 - 5 | 618 | 106 | |
AY061886.1 | - | 4 - 5 | 525 | 106 | |
BC113505.1 | - | 4 - 5 | 603 | 106 | |
BC113507.1 | - | 4 - 5 | 603 | 106 |