The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001257378.1 | NP_001244307.1 | 3 - 4 | 270 | 75 |
NM_001257379.1 | NP_001244308.1 | 3 - 4 | 270 | 75 | |
NM_001257382.1 | NP_001244311.1 | 3 - 4 | 340 | 75 | |
NM_174975.4 | NP_777635.1 | 2 - 3 | 222 | 75 | |
XM_011530128.2 | XP_011528430.1 | 2 - 3 | 190 | 75 | |
GenBank mRNA | AK131358.1 | - | 3 - 4 | 244 | 75 |
AK298079.1 | - | 3 - 4 | 214 | 75 | |
AY158086.1 | - | 2 - 3 | 133 | 75 | |
AY240872.1 | - | 2 - 3 | 222 | 75 | |
BC101001.2 | - | 3 - 4 | 209 | 75 | |
BC101002.1 | - | 3 - 4 | 209 | 75 | |
BC101003.1 | - | 3 - 4 | 279 | 75 | |
BC101004.1 | - | 2 - 3 | 161 | 75 | |
BU584962.1 | - | 2 - 3 | 143 | 75 |