The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001257378.1 | NP_001244307.1 | 8 - 9 | 827 | 74 |
NM_001257379.1 | NP_001244308.1 | 7 - 8 | 657 | 74 | |
NM_001257382.1 | NP_001244311.1 | 7 - 8 | 727 | 74 | |
NM_174975.4 | NP_777635.1 | 6 - 7 | 609 | 74 | |
XM_011530128.2 | XP_011528430.1 | 6 - 7 | 577 | 74 | |
GenBank mRNA | AK131358.1 | - | 7 - 8 | 631 | 74 |
AY158086.1 | - | 6 - 7 | 520 | 74 | |
AY240872.1 | - | 6 - 7 | 609 | 74 | |
BC069641.1 | - | 6 - 7 | 532 | 74 | |
BC101001.2 | - | 8 - 9 | 766 | 74 | |
BC101002.1 | - | 7 - 8 | 596 | 74 | |
BC101003.1 | - | 7 - 8 | 666 | 74 | |
BC101004.1 | - | 6 - 7 | 548 | 74 |