Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
POTED
Entrez Gene ID
317754
Gene Name
POTE ankyrin domain family member D
Gene Aliases
A26B3, ANKRD21, CT104.1, POTE, POTE-21, POTE21
Chromosome Location
Chr.21: 13610081 - 13641585 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_174981.3 NP_778146.2 1 - 2 569 100
XM_006723997.3 XP_006724060.1 1 - 2 665 100
XM_011529550.2 XP_011527852.1 1 - 2 665 100

Gene Symbol
LOC100288966
Entrez Gene ID
100288966
Gene Name
POTE ankyrin domain family member D-like
Gene Aliases
-
Chromosome Location
Chr.Un|NT_167208: 108007 - 139435 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001257362.1 NP_001244291.1 1 - 2 569 100
XM_006709930.3 XP_006709993.1 1 - 2 665 100
XM_011545655.2 XP_011543957.1 1 - 2 665 100
XM_011545656.2 XP_011543958.1 1 - 2 665 100
GenBank mRNA AY172978.1 - 1 - 2 569 100

View Full Product Details